Pierce™ Protein A IgG Plus Orientation Kit, 2 mL
Thermo Scientific™

Pierce™ Protein A IgG Plus Orientation Kit, 2 mL

The Thermo Scientific Pierce A IgG Plus Orientation Kit uses a crosslinker to covalently attach antibody that is bound toRead more
Have Questions?
Catalog NumberQuantity
448932 Columns
Catalog number 44893
Price (USD)
872.00
Each
Quantity:
2 Columns
Request bulk or custom format
Price (USD)
872.00
Each
The Thermo Scientific Pierce A IgG Plus Orientation Kit uses a crosslinker to covalently attach antibody that is bound to Protein A agarose resin, thereby creating a reusable affinity column for purification of antigens.

Features of the Protein A IgG Plus Orientation Kit:

• Contaminant-free antigen purification—provides for antigen purification without contamination (co-elution) of antibody
• Covalent, leak-resistant antibody immobilization—high-capacity binding of IgG to Protein A agarose, followed by efficient crosslinking with DSS crosslinker
• High-quality agarose resin—crosslinked 6% beaded agarose with recombinant Protein A
• Gentle binding and crosslinking conditions—coupling antibody in standard PBS
• Oriented immobilization—binds and crosslinks IgG with the antigen-binding sites directed outward, able to capture passing antigen in the mobile phase without steric hindrance
• Complete kit—includes all the reagents needed to immobilize the antibody
• Better than DMP—less antibody leaching than traditional orientation method with DMP crosslinker

This 'Plus' kit provides for more efficient and stable antibody immobilization than the traditional orientation method because it uses the NHS-ester crosslinker DSS (disuccinimidyl suberate) rather than the imidoester crosslinker DMP (dimethyl pimelimidate). Several milligrams of IgG can be bound and crosslinked to each 2 ml column, after which the prepared affinity column can be used multiple times for antigen purification using typical binding and elution conditions.

Related Products
Pierce™ Protein G IgG Plus Orientation Kit, 2 mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)2 mL
Column TypeAffinity, Agarose Resin
FormatSpin Column
Quantity2 Columns
Stationary PhaseProtein A
Sufficient For2 to 10 mg of Rabbit IgG per Column
Product LinePierce
TypeIgG Plus Orientation Kit
Unit SizeEach
Contents & Storage
Sufficient For: Immobilizing 2 to 10 mg of rabbit IgG per column
• Recombinant Protein A Columns, 2 mL, 2 columns
• Disuccinimidyl Suberate (DSS), 2 x 13 mg
• PBS Pack (makes 500 mL), 1 pack
• Blocking Buffer, 6 mL
• IgG Elution Buffer, 15 mL
• Antibody Binding/Wash Buffer, 120 mL
• Column Accessories

Store at 4°C.

Frequently asked questions (FAQs)

What are the Pierce Protein A and Protein G IgG Plus Orientation kits for?

The Orientation kits allow for binding of an antibody to the Protein A or G resin. After the antibody is bound, an amine- reactive crosslinker (DSS) is added to covalently attach the antibody to the support. This allows for the target antigen to elute free of antibody contamination. The Protein A and G Orientation Kits are used for larger-scale purification of IgG than is typical in an immunoprecipitation.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.