Lithium chloride, 99%, for molecular biology - FAQs
Lithium chloride, 99%, for molecular biology - FAQs
View additional product information for Lithium chloride, 99%, for molecular biology - FAQs (449045000, 449041000)
1 product FAQs found
What storage conditions should I use for Lithium chloride?
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.