Prednisolone, 99%, Thermo Scientific Chemicals
Prednisolone, 99%, Thermo Scientific Chemicals
Prednisolone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Prednisolone, 99%, Thermo Scientific Chemicals

Prednisolone, or metacortandralone, CAS # 50-24-8, is a synthetic glucocorticoid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
4494700101gGlass bottle
4494700505gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 449470010
Price (USD)
42.30
Each
Quantity:
1g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.30
Each
Chemical Identifiers
CAS50-24-8
IUPAC Name(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular FormulaC21H28O5
InChI KeyOIGNJSKKLXVSLS-VWUMJDOOSA-N
SMILESC[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Appearance (Color)White to off-white
Specific optical rotation+113° to +119° (20°C, 589 nm) (c=1, ethanol)
Appearance (Form)Powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Prednisolone is a derivative of prednisone with an oxo to beta-hydroxy reduction at position 11

• Prednisolone binds cell surface receptors and is transported to the nucleus, where it binds certain nuclear receptors responsible for transcription of proinflammatory cytokines

Applications

• Prednisolone can be used for laboratory research on tumor growth and prevention

• Prednisolone may also be used for developmental and endocrine research, in vivo or in vitro

RUO – Research Use Only

Literature References

Waldrop, M.A.; Karingada, C.; Storey, M.A.; Powers, B.; Lammarino, M.A.; Miller, N.F.; Alfano, L.N.; Noritz, G.; Rossman, I.; Ginsberg, M.; Mosher, K.A., Broomall, E.; Goldstein, J.; Bass, N.; Lowes, L.P.; Tsao, C.Y.; Mendell, J.R.; Connolly, A.M. Gene Therapy for Spinal Muscular Atrophy: Safety and Early Outcomes. Pediatrics. 2020, 146(3), e20200729.

Koumpa, F.S.; Forde, C.T.; Manjaly, J.G. Sudden irreversible hearing loss post COVID-19. BMJ Case Rep. 2020, 13(11), e238419.