Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Thermo Scientific Chemicals

Cytidine 5'-monophosphate, 97%, from yeast

CAS: 63-37-6 | C9H14N3O8P | 323.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 449490010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS63-37-6
IUPAC Name{[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Molecular FormulaC9H14N3O8P
InChI KeyIERHLVCPSMICTF-NHBCCFCANA-N
SMILESNC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white
HPLC>=96.0 %
Infrared spectrumConforms
Loss on drying=<3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only