2-Quinolinecarbonitrile, 97%
2-Quinolinecarbonitrile, 97%
2-Quinolinecarbonitrile, 97%
Thermo Scientific Chemicals

2-Quinolinecarbonitrile, 97%

CAS: 1436-43-7 | C10H6N2 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 449580010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1436-43-7
IUPAC Namequinoline-2-carbonitrile
Molecular FormulaC10H6N2
InChI KeyWDXARTMCIRVMAE-UHFFFAOYSA-N
SMILESN#CC1=CC=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point92°C to 96°C
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only