2-Chloro-4-ethynylpyridine, 97%
2-Chloro-4-ethynylpyridine, 97%
2-Chloro-4-ethynylpyridine, 97%
Thermo Scientific Chemicals

2-Chloro-4-ethynylpyridine, 97%

CAS: 945717-09-9 | C7H4ClN | 137.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 449980050
Price (USD)
830.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
830.00
Each
Chemical Identifiers
CAS945717-09-9
IUPAC Name2-chloro-4-ethynylpyridine
Molecular FormulaC7H4ClN
InChI KeyDLEWWBKXDRBGRF-UHFFFAOYSA-N
SMILESClC1=NC=CC(=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light brown to green
HPLC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only