Search
View additional product information for Human beta-NGF Recombinant Protein, PeproTech® - FAQs (450-01-20UG, 450-01-250UG, 450-01-500UG, 450-01-1MG, 450-01-100UG)
1 product FAQs found
For the PeproTech products "lyophilized with no additives" it means that the solvents present during the lyophilization process evaporate out. For most of these products, we generally recommend using water for the initial reconstitution. However, this is not always the case. Therefore, we advise referring to the lot-specific Certificate of Analysis (CoA) for the most accurate reconstitution instructions for each vial.
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .