Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 85,000 to 120,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 85,000 to 120,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 85,000 to 120,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 85,000 to 120,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
45014025025gPlastic bottle
450145000500gPlastic bottle
4501400101kgPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 450140250
Price (EUR)
25,90
Each
Quantity:
25g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
25,90
Each
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Degree of hydrolysis87.0 to 89.0 mol %
pH4.5 to 6.5 (4 % in water)
Ash=<0.5 % (Na2O)
Appearance (Color)White to off-white
Viscosity23.0 to 27.0 cP (4 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only