Metaphosphoric acid, ACS reagent, stabilized
Metaphosphoric acid, ACS reagent, stabilized
Metaphosphoric acid, ACS reagent, stabilized
Thermo Scientific Chemicals

Metaphosphoric acid, ACS reagent, stabilized

CAS: 37267-86-0 | HO3P | 79.979 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Glass Bottle
製品番号(カタログ番号) 450255000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS37267-86-0
IUPAC Nametrihydroxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Molecular FormulaH3O9P3
InChI KeyAZSFNUJOCKMOGB-UHFFFAOYSA-N
SMILESOP1(=O)OP(O)(=O)OP(O)(=O)O1
さらに表示
Titration with NaOH33.5 to 36.5 % (HPO3)
Nitrate (NO3)=<0.001 %
Arsenic (As)=<1 ppm
KMnO4-reducing substance=<0.02 % (as H3PO3)
Appearance (Color)Colorless to white
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only