3,5-Diiodosalicylaldehyde, 97%
3,5-Diiodosalicylaldehyde, 97%
3,5-Diiodosalicylaldehyde, 97%
Thermo Scientific Chemicals

3,5-Diiodosalicylaldehyde, 97%

CAS: 2631-77-8 | C7H4I2O2 | 373.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 450860050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2631-77-8
IUPAC Name2-hydroxy-3,5-diiodobenzaldehyde
Molecular FormulaC7H4I2O2
InChI KeyMYWSBJKVOUZCIA-UHFFFAOYSA-N
SMILESOC1=C(I)C=C(I)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
Melting point107°C to 112°C
Appearance (Color)Yellow to light brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only