2,2,3,3-Tetramethoxybutane, 95%
2,2,3,3-Tetramethoxybutane, 95%
2,2,3,3-Tetramethoxybutane, 95%
Thermo Scientific Chemicals

2,2,3,3-Tetramethoxybutane, 95%

CAS: 176798-33-7 | C8H18O4 | 178.23 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 450930250
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS176798-33-7
IUPAC Name2,2,3,3-tetramethoxybutane
Molecular FormulaC8H18O4
InChI KeyCOEBCOIFCLAOFZ-UHFFFAOYSA-N
SMILESCOC(C)(OC)C(C)(OC)OC
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only