Yeast extract
Thermo Scientific Chemicals

Yeast extract

CAS: 84604-16-
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5kg
25 kg
Catalog number 451120250
Price (USD)
834.65
Online exclusive
927.00
Save 92.35
Each
Quantity:
25 kg
Request bulk or custom format
Price (USD)
834.65
Online exclusive
927.00
Save 92.35
Each
Chemical Identifiers
CAS84604-16-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total nitrogen>=9.0 % (on dry substance)
Amino nitrogen>=3.0 % (on dry substance)
Loss on drying=<6.0 %
pH5.3 to 7.2 (2 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only