When using the Zero Blunt TOPO PCR Cloning Kit (Cat. No. 450245), what is the recommended ratio of vector to insert?
The recommended ratio of vector:insert is 1:1 to 1:3.
Find additional tips, troubleshooting help, and resources within our Cloning Support Center.
Find additional tips, troubleshooting help, and resources within our Cloning Support Center.