Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4514800505 g
Catalog number 451480050
Price (USD)
245.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
245.00
Each
Chemical Identifiers
CAS42899-76-3
IUPAC Namepyridine-3-sulfonyl chloride hydrochloride
Molecular FormulaC5H5Cl2NO2S
InChI KeyVIVPWOMJFLICOZ-UHFFFAOYSA-N
SMILESCl.ClS(=O)(=O)C1=CN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder or solid
Infrared spectrumConforms
Titration Argentometric94.0 to 106.0 %
Elemental Analysis26.37 to 29.74 % (C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only