4-Bromobenzylsulfonyl chloride, 95%, Thermo Scientific Chemicals
4-Bromobenzylsulfonyl chloride, 95%, Thermo Scientific Chemicals
4-Bromobenzylsulfonyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromobenzylsulfonyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4514900505 g
Catalog number 451490050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS53531-69-4
IUPAC Name(4-bromophenyl)methanesulfonyl chloride
Molecular FormulaC7H6BrClO2S
InChI KeyZWVWFWGJZPHCHF-UHFFFAOYSA-N
SMILESClS(=O)(=O)CC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Color)White to off-white
Appearance (Form)Powder or solid
Melting point116°C to 121°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only