Propargyl chloride, 98%
Propargyl chloride, 98%
Propargyl chloride, 98%
Thermo Scientific Chemicals

Propargyl chloride, 98%

CAS: 624-65-7 | C3H3Cl | 74.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 451840250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS624-65-7
IUPAC Name3-chloroprop-1-yne
Molecular FormulaC3H3Cl
InChI KeyLJZPPWWHKPGCHS-UHFFFAOYSA-N
SMILESClCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Colorless to yellow to light brown
Infrared spectrumConforms
Refractive index1.4340 to 1.4370 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only