Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 97%
Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 97%
Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 97%

CAS: 155377-19-8 | C7H7F3N2O2 | 208.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 452280050
Price (USD)
135.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
135.00
Each
Chemical Identifiers
CAS155377-19-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point139°C to 145°C
Appearance (Color)Off-white to yellow to tan
Appearance (Form)Powder or solid
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only