3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%
3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%
3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%
Thermo Scientific Chemicals

3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%

CAS: 175277-11-9 | C9H14N2O2 | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number 452620050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS175277-11-9
IUPAC Name3-tert-butyl-1-methyl-1H-pyrazole-5-carboxylic acid
Molecular FormulaC9H14N2O2
InChI KeySFSXXMXHJOSBAZ-UHFFFAOYSA-N
SMILESCN1N=C(C=C1C(O)=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Melting point150°C to 156°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only