Methyl Arachidate, 99%
Methyl Arachidate, 99%
Methyl Arachidate, 99%
Thermo Scientific Chemicals

Methyl Arachidate, 99%

CAS: 1120-28-1 | C21H42O2 | 326.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 452820010
Price (USD)
89.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
89.10
Each
Chemical Identifiers
CAS1120-28-1
IUPAC Namemethyl icosanoate
Molecular FormulaC21H42O2
InChI KeyQGBRLVONZXHAKJ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Free acid=<1 %
Appearance (Color)White to almost white
Appearance (Form)Powder or flakes
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only