Tetrahydropyran-2-ylmethylamine, 97%
Tetrahydropyran-2-ylmethylamine, 97%
Tetrahydropyran-2-ylmethylamine, 97%
Thermo Scientific Chemicals

Tetrahydropyran-2-ylmethylamine, 97%

CAS: 6628-83-7 | C6H13NO | 115.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 453370010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS6628-83-7
IUPAC Name1-(oxan-2-yl)methanamine
Molecular FormulaC6H13NO
InChI KeyNYGCBQKDTGBHSC-UHFFFAOYNA-N
SMILESNCC1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only