Barium nitrate, pure
Barium nitrate, pure
Barium nitrate, pure
Thermo Scientific Chemicals

Barium nitrate, pure

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Catalog number 453800025
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Chemical Identifiers
CAS10022-31-8
IUPAC Namebarium(2+) dinitrate
Molecular FormulaBaN2O6
InChI KeyIWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric>=98.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only