Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals
Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals
Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
453851000100 g
Catalog number 453851000
Price (EUR)
77,65
Online exclusive
85,90
Save 8,25 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
77,65
Online exclusive
85,90
Save 8,25 (10%)
Each
Chemical Identifiers
CAS7787-68-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Assay>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only