Sulfuro de bismuto(III), 99 %, puro, Thermo Scientific Chemicals
Sulfuro de bismuto(III), 99 %, puro, Thermo Scientific Chemicals
Sulfuro de bismuto(III), 99 %, puro, Thermo Scientific Chemicals
Sulfuro de bismuto(III), 99 %, puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfuro de bismuto(III), 99 %, puro, Thermo Scientific Chemicals

CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
Número de catálogo 453995000
Precio (USD)
-
Cantidad:
500 g
Pedido a granel o personalizado
Identificadores químicos
CAS1345-07-9
IUPAC Namedibismuth(3+) trisulfanediide
Molecular FormulaBi2S3
InChI KeyYNRGZHRFBQOYPP-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Bi+3].[Bi+3]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Titration Complexometric80.1 to 82.5 % (Bi)
Appearance (Form)Powder and/or chunks
Appearance (Color)Grey

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only