5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific Chemicals
5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific Chemicals
5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific Chemicals

CAS: 57489-77-7 | C6H3Cl2N3 | 188.02 g/mol
Have Questions?
Catalog NumberQuantity
4541300101 g
Catalog number 454130010
Price (USD)
483.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
483.00
Each
Chemical Identifiers
CAS57489-77-7
IUPAC Name5,7-dichloropyrazolo[1,5-a]pyrimidine
Molecular FormulaC6H3Cl2N3
InChI KeyJMTFWCYVZOFHLR-UHFFFAOYSA-N
SMILESClC1=NC2=CC=NN2C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)White to yellow
Appearance (Form)Solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only