2,3,5-Trimethylfuran, 99%
2,3,5-Trimethylfuran, 99%
2,3,5-Trimethylfuran, 99%
Thermo Scientific Chemicals

2,3,5-Trimethylfuran, 99%

CAS: 10504-04-8 | C7H10O | 110.16 g/mol
Have Questions?
Catalog NumberQuantity
4544700101 g
Catalog number 454470010
Price (USD)
116.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
116.00
Each
Chemical Identifiers
CAS10504-04-8
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only