2-Aminomethyl-1H-imidazole dihydrochloride, 97%
2-Aminomethyl-1H-imidazole dihydrochloride, 97%
2-Aminomethyl-1H-imidazole dihydrochloride, 97%
Thermo Scientific Chemicals

2-Aminomethyl-1H-imidazole dihydrochloride, 97%

CAS: 22600-77-7 | C4H9Cl2N3 | 170.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 454590050
Price (USD)
276.65
Online exclusive
307.00
Save 30.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
276.65
Online exclusive
307.00
Save 30.35
Each
Chemical Identifiers
CAS22600-77-7
IUPAC Name1-(1H-imidazol-2-yl)methanamine dihydrochloride
Molecular FormulaC4H9Cl2N3
InChI KeyKYUDBQDDNKPSIC-UHFFFAOYSA-N
SMILESCl.Cl.NCC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
HPLC>=96.0 %
Titration Argentometric96.0 to 104.0 %
Water=<5 %
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only