2-Bromoanthracene, 98%, Thermo Scientific Chemicals
2-Bromoanthracene, 98%, Thermo Scientific Chemicals
2-Bromoanthracene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoanthracene, 98%, Thermo Scientific Chemicals

CAS: 7321-27-9 | C14H9Br | 257.13 g/mol
Have Questions?
Catalog NumberQuantity
4548800101 g
Catalog number 454880010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7321-27-9
IUPAC Name2-bromoanthracene
Molecular FormulaC14H9Br
InChI KeyPYXBCVWIECUMDW-UHFFFAOYSA-N
SMILESBrC1=CC2=CC3=CC=CC=C3C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light brown
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only