4,4'-Dibromo-2,2'-bipyridine, 98%
4,4'-Dibromo-2,2'-bipyridine, 98%
4,4'-Dibromo-2,2'-bipyridine, 98%
Thermo Scientific Chemicals

4,4'-Dibromo-2,2'-bipyridine, 98%

CAS: 18511-71-2 | C10H6Br2N2 | 313.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 454900010
Price (USD)
107.65
Special offer
Online exclusive
Ends: 28-Dec-2026
126.00
Save 18.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 28-Dec-2026
126.00
Save 18.35
Each
Chemical Identifiers
CAS18511-71-2
IUPAC Name4,4'-dibromo-2,2'-bipyridine
Molecular FormulaC10H6Br2N2
InChI KeyKIIHBDSNVJRWFY-UHFFFAOYSA-N
SMILESBrC1=CC(=NC=C1)C1=NC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point140°C to 145°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only