3'-Deoxy-2',3'-didehydrothymidine, 98%
3'-Deoxy-2',3'-didehydrothymidine, 98%
3'-Deoxy-2',3'-didehydrothymidine, 98%
Thermo Scientific Chemicals

3'-Deoxy-2',3'-didehydrothymidine, 98%

CAS: 3056-17-5 | C10H12N2O4 | 224.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 454940010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3056-17-5
IUPAC Name1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC10H12N2O4
InChI KeyXNKLLVCARDGLGL-HGXVMFPFNA-N
SMILESCC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Specific optical rotation-44.0° to -48.0° (20°C, 589 nm) (c=0.7, water)
Melting point160°C to 169°C
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only