Ethyl viologen dibromide, 97%
Ethyl viologen dibromide, 97%
Ethyl viologen dibromide, 97%
Thermo Scientific Chemicals

Ethyl viologen dibromide, 97%

CAS: 53721-12-3 | C14H18Br2N2 | 374.11 g/mol
製品番号(カタログ番号) 455030050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS53721-12-3
IUPAC Name1,1'-diethyl-[4,4'-bipyridine]-1,1'-diium dibromide
Molecular FormulaC14H18Br2N2
InChI KeyLCEBDKLPALDQPV-UHFFFAOYSA-L
SMILES[Br-].[Br-].CC[N+]1=CC=C(C=C1)C1=CC=[N+](CC)C=C1
さらに表示
Appearance (Form)Crystalline powder
Titration Argentometric>=96.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only