Amoxicillin, 96%
Amoxicillin, 96%
Amoxicillin, 96%
Thermo Scientific Chemicals

Amoxicillin, 96%

CAS: 26787-78-0 | C16H19N3O5S | 365.404 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 455140050
Price (EUR)
77,65
Online exclusive
86,40
Save 8,75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
77,65
Online exclusive
86,40
Save 8,75 (10%)
Each
Specifications
CAS26787-78-0
ChEBICHEBI:2676
Chemical Name or MaterialAmoxicillin
Formula Weight365.4
Health Hazard 1Danger
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only