Amphotericin B, 85%
Amphotericin B, 85%
Amphotericin B, 85%
Thermo Scientific Chemicals

Amphotericin B, 85%

Amphotericin B, CAS # 1397-89-3, is an antifungal that has been used against fungus caused by protozoan parasites of Leishmania genus. | CAS: 1397-89-3 | C47H73NO17 | 924.09 g/mol
Have Questions?
Catalog NumberQuantity
4554900101 g
Catalog number 455490010
Price (USD)
164.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
164.00
Each
Chemical Identifiers
CAS1397-89-3
IUPAC Name(1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
Molecular FormulaC47H73NO17
InChI KeyAPKFDSVGJQXUKY-ZNVUZQDLSA-N
SMILESC[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV spectrumComplies
HPLC>=80 %
Water=<10 %
Appearance (Color)Yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Amphotericin B is an antibiotic that has antifungal activity and is produced mainly by Streptomyces nodosus

Applications

• It has antifungal activity against fungus caused by protozoan parasites of Leishmania genus by binding to an essential component of the fungal cell membrane and altering cell membrane permeability

• It can suppress bone marrow progenitor cells and induce Actinomycin D(CR001) sensitivity in drug resistant HELA cells in vitro

RUO – Research Use Only

General References:

  1. Boulware, R.D. et. al. Timing of antiretroviral therapy after diagnosis of cryptococcal meningitis. New England Journal of Medicine. 2014, 370, (26), 2487-2498.
  2. Sarkar, S. et. al. Therapeutic activation of macrophages and microglia to suppress brain tumor-initiating cells. Nature Neuroscience. 2014, 17, (1), 46-55.