2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals
2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals
2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals

CAS: 15103-48-7 | Molecular Formula: C5H5NO3S | Molecular Weight: 159.16 g/mol
Have Questions?
Catalog NumberQuantity
4557600505 g
Catalog number 455760050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS15103-48-7
IUPAC Namepyridine-2-sulfonic acid
Molecular FormulaC5H5NO3S
InChI KeyKZVLNAGYSAKYMG-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Melting point245°C to 252°C
HPLC>=96.0 %
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only