p-Toluenesulfonyl cyanide, 95%
p-Toluenesulfonyl cyanide, 95%
p-Toluenesulfonyl cyanide, 95%
Thermo Scientific Chemicals

p-Toluenesulfonyl cyanide, 95%

CAS: 19158-51-1 | C8H7NO2S | 181.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 455820010
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.50
Save 10.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.50
Save 10.85
Each
Chemical Identifiers
CAS19158-51-1
IUPAC Name4-methylbenzene-1-sulfonyl cyanide
Molecular FormulaC8H7NO2S
InChI KeyJONIMGVUGJVFQD-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point47°C to 50°C
Appearance (Color)White to tan
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only