Oxaliplatin, 99%
Oxaliplatin, 99%
Oxaliplatin, 99%
Thermo Scientific Chemicals

Oxaliplatin, 99%

CAS: 61825-94-3 | C8H14N2O4Pt | 397.29 g/mol
Have Questions?
Catalog NumberQuantity
456131000100 mg
Catalog number 456131000
Price (USD)
205.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
205.00
Each
Chemical Identifiers
CAS61825-94-3
IUPAC Name(1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
Molecular FormulaC8H14N2O4Pt
InChI KeyZROHGHOFXNOHSO-WHMNLBETNA-L
SMILESO=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying=<0.5 % (1 g, 105°C, 2 h)
Appearance (Form)Powder
HPLC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only