Topiramate, 98%
Topiramate, 98%
Topiramate, 98%
Thermo Scientific Chemicals

Topiramate, 98%

CAS: 97240-79-4 | C12H21NO8S | 339.36 g/mol
Have Questions?
Catalog NumberQuantity
4561900101 g
Catalog number 456190010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS97240-79-4
IUPAC Name[(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-6-yl]methyl sulfamate
Molecular FormulaC12H21NO8S
InChI KeyKJADKKWYZYXHBB-XBWDGYHZSA-N
SMILESCC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point124°C to 128°C
Specific optical rotation-29° to -33° (20°C, 589 nm) (c=1, methanol)
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only