Forchlorfenuron, 98%
Forchlorfenuron, 98%
Forchlorfenuron, 98%
Forchlorfenuron, 98%
Thermo Scientific Chemicals

Forchlorfenuron, 98%

CAS: 68157-60-8 | C12H10ClN3O | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 456290010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS68157-60-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<1 %
Appearance (Color)White to light yellow
HPLC>=97.5 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only