1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct, 97%
1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct, 97%
1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct, 97%
Thermo Scientific Chemicals

1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct, 97%

CAS: 119752-83-9 | C6H12N2O4S2 | 240.3 g/mol
Have Questions?
Catalog NumberQuantity
4563500101 g
Catalog number 456350010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS119752-83-9
IUPAC Name1,4-diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulfinate
Molecular FormulaC6H12N2O4S2
InChI KeyRWISEVUOFYXWFO-UHFFFAOYSA-N
SMILES[O-]S(=O)[N+]12CC[N+](CC1)(CC2)S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=96.0 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point168°C to 172°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only