N-Formyl-L-proline, 97%
N-Formyl-L-proline, 97%
N-Formyl-L-proline, 97%
Thermo Scientific Chemicals

N-Formyl-L-proline, 97%

CAS: 13200-83-4 | C6H9NO3 | 143.14 g/mol
製品番号(カタログ番号) 456420010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13200-83-4
IUPAC Name1-formylpyrrolidine-2-carboxylic acid
Molecular FormulaC6H9NO3
InChI KeyDHDRGOURKDLAOT-UHFFFAOYNA-N
SMILESOC(=O)C1CCCN1C=O
さらに表示
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
Specific optical rotation-115° to -130° (20°C, 589 nm) (c=0.8 in EtOH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only