Montelukast sodium, 98%
Montelukast sodium, 98%
Montelukast sodium, 98%
Thermo Scientific Chemicals

Montelukast sodium, 98%

CAS: 151767-02-1 | C35H35ClNNaO3S | 608.17 g/mol
Have Questions?
Catalog NumberQuantity
4564400101 g
Catalog number 456440010
Price (USD)
161.65
Special offer
Online exclusive
Ends: 28-Dec-2026
190.00
Save 28.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
161.65
Special offer
Online exclusive
Ends: 28-Dec-2026
190.00
Save 28.35
Each
Chemical Identifiers
CAS151767-02-1
IUPAC Namesodium 2-[1-({[(1R)-1-{3-[(1E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl}-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanyl}methyl)cyclopropyl]acetate
Molecular FormulaC35H35ClNNaO3S
InChI KeyLBFBRXGCXUHRJY-HKHDRNBDSA-M
SMILES[Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
HPLC>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Water=<2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only