(R)-3-Methylmorpholine hydrochloride, 97%
(R)-3-Methylmorpholine hydrochloride, 97%
(R)-3-Methylmorpholine hydrochloride, 97%
Thermo Scientific Chemicals

(R)-3-Methylmorpholine hydrochloride, 97%

CAS: 953780-78-4 | C5H11NO·ClH | 137.61 g/mol
Have Questions?
Catalog NumberQuantity
4567800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 456780010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS953780-78-4
IUPAC Name3-methylmorpholine hydrochloride
Molecular FormulaC5H12ClNO
InChI KeyMSOCQCWIEBVSLF-UHFFFAOYNA-N
SMILESCl.CC1COCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
Titration Argentometric>=96.0 to =<104.0 %
Specific optical rotation+1° to +3° (20°C, 589 nm) c=1 in H2O
Appearance (Form)Powder or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only