Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Thermo Scientific™

Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 457071000
Price (USD)
31.65
Online Exclusive
35.00
Save 3.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
35.00
Save 3.35 (10%)
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Peroxides=<0.0075 %
Residue after evaporation=<0.001 %
Appearance (Form)Clear liquid
GC>=99.80 %
Water=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only