3-Acetylpyridine adenine dinucleotide, 90%
3-Acetylpyridine adenine dinucleotide, 90%
3-Acetylpyridine adenine dinucleotide, 90%
Thermo Scientific Chemicals

3-Acetylpyridine adenine dinucleotide, 90%

CAS: 86-08-8 | C22H28N6O14P2 | 662.44 g/mol
Have Questions?
Catalog NumberQuantity
4576800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 457680010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS86-08-8
IUPAC Name3-acetyl-1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium
Molecular FormulaC22H28N6O14P2
InChI KeyKPVQNXLUPNWQHM-RBEMOOQDSA-N
SMILESCC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=89.0 %
Water=<8 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only