Iodine, specified according to the requirements of Ph.Eur, BP, USP
Iodine, specified according to the requirements of Ph.Eur, BP, USP
Iodine, specified according to the requirements of Ph.Eur, BP, USP
Thermo Scientific Chemicals

Iodine, specified according to the requirements of Ph.Eur, BP, USP

CAS: 7553-56-2 | I2 | 253.81 g/mol
Quantity:
250 g
1 kg
Catalog number 457762500
Price (JPY)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Greyish-violet
Appearance (Form)Brittle plates or crystals with metallic sheen
Non-volatile matter=<0.05 %
Titration Iodometric99.8 to 100.5 %
Chloride and Bromide=<250 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only