Aripiprazole
Aripiprazole
Aripiprazole
Thermo Scientific Chemicals

Aripiprazole

CAS: 129722-12-9 | C23H27Cl2N3O2 | 448.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 457990050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS129722-12-9
IUPAC Name7-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy}-1,2,3,4-tetrahydroquinolin-2-one
Molecular FormulaC23H27Cl2N3O2
InChI KeyCEUORZQYGODEFX-UHFFFAOYSA-N
SMILESClC1=CC=CC(N2CCN(CCCCOC3=CC=C4CCC(=O)NC4=C3)CC2)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Melting point138°C to 150°C
Appearance (Color)White to beige
Infrared spectrumConforms
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only