1-Bromo-3,4,5-trimethoxybenzene, 97%
1-Bromo-3,4,5-trimethoxybenzene, 97%
1-Bromo-3,4,5-trimethoxybenzene, 97%
Thermo Scientific Chemicals

1-Bromo-3,4,5-trimethoxybenzene, 97%

CAS: 2675-79-8 | C9H11BrO3 | 247.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 458130050
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.70
Save 6.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.70
Save 6.05
Each
Chemical Identifiers
CAS2675-79-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point75°C to 83°C
Appearance (Color)White to brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only