2-Bromopyrazine, 95%
2-Bromopyrazine, 95%
2-Bromopyrazine, 95%
Thermo Scientific Chemicals

2-Bromopyrazine, 95%

CAS: 56423-63-3 | C4H3BrN2 | 159.99 g/mol
Have Questions?
Catalog NumberQuantity
4581600101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458160010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS56423-63-3
IUPAC Name2-bromopyrazine
Molecular FormulaC4H3BrN2
InChI KeyWGFCNCNTGOFBBF-UHFFFAOYSA-N
SMILESBrC1=NC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94.0 %
Appearance (Color)Clear colorless to light orange
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only