TCF-32 Continuous-Flow Rotors
Thermo Scientific™
TCF-32 Continuous-Flow Rotors
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of these rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityDescription
45820940mL/430 mLTCF-32 Continuous-Flow Rotor
45830940 mL940mL Accessory Core
45835430mL430mL Standard Core
Catalog number 45820
Price (ZAR)
-
Capacity:
940mL/430 mL
Description:
TCF-32 Continuous-Flow Rotor
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of Thermo Scientific™ TCF-32 Continuous-Flow Rotors.
Compatible with:
Thermo Scientific Sorvall WX Ultra Centrifuge Series
Ordering Information:
Core is required for operation. Must be purchased separately.
Specifications
AutoclavableAutoclavable
DescriptionTCF-32 Continuous-Flow Rotor
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
K Factor106/42
Capacity940mL/430 mL
Max. RCF102,000 x g
Max. Speed32,000 rpm
TypeContinuous Flow Rotor
Unit SizeEach