3,4-Dihydro-2H-isoquinolin-1-one, 98%
3,4-Dihydro-2H-isoquinolin-1-one, 98%
3,4-Dihydro-2H-isoquinolin-1-one, 98%
Thermo Scientific Chemicals

3,4-Dihydro-2H-isoquinolin-1-one, 98%

CAS: 1196-38-9 | C9H9NO | 147.18 g/mol
Quantity:
1 g
5 g
Catalog number 458220010
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1196-38-9
IUPAC Name1,2,3,4-tetrahydroisoquinolin-1-one
Molecular FormulaC9H9NO
InChI KeyYWPMKTWUFVOFPL-UHFFFAOYSA-N
SMILESO=C1NCCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only