Fusidic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fusidic acid, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
4584600505g
製品番号(カタログ番号) 458460050
価格(JPY)
-
数量:
5g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6990-06-3
IUPAC Name2-[(1Z,2S,3aS,3bS,5aS,6S,7R,9aS,9bS,10R,11aR)-2-(acetyloxy)-7,10-dihydroxy-3a,3b,6,9a-tetramethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-ylidene]-6-methylhept-5-enoic acid
Molecular FormulaC31H48O6
InChI KeyIECPWNUMDGFDKC-MZJAQBGESA-N
SMILESC[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]1(C)[C@H]2[C@H](O)C[C@H]2C([C@H](C[C@]12C)OC(C)=O)=C(/CCC=C(C)C)C(O)=O
さらに表示
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Water=<10 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only